Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 6 - 7 | 882 | 72 | |
| GenBank mRNA | AK044203.1 | - | 6 - 7 | 855 | 72 | |
| AK140375.1 | - | 6 - 7 | 855 | 72 | ||
| BC012875.1 | - | 6 - 7 | 802 | 3500318 | 72 | |
| CT010331.1 | - | 5 - 6 | 733 | 72 | ||
More Information
Set Membership: |
|

Anxa11
Genomic Map