Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 8 - 9 | 1084 | 64 | |
| GenBank mRNA | AK044203.1 | - | 8 - 9 | 1057 | 64 | |
| AK137158.1 | - | 7 - 8 | 1394 | 64 | ||
| AK140375.1 | - | 8 - 9 | 1057 | 64 | ||
| AK170850.1 | - | 6 - 7 | 2468 | 64 | ||
| BC012875.1 | - | 8 - 9 | 1004 | 3500318 | 64 | |
| CT010331.1 | - | 7 - 8 | 935 | 64 | ||
| U65986.1 | - | 8 - 9 | 1069 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Anxa11
Genomic Map