Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 11 - 12 | 1320 | 80 | |
| GenBank mRNA | AK137158.1 | - | 10 - 11 | 1630 | 80 | |
| AK140375.1 | - | 11 - 12 | 1293 | 80 | ||
| BC012875.1 | - | 11 - 12 | 1240 | 3500318 | 80 | |
| CT010331.1 | - | 10 - 11 | 1171 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa11
Genomic Map