Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 12 - 13 | 1421 | 132 | |
| GenBank mRNA | AK137158.1 | - | 11 - 12 | 1731 | 132 | |
| AK140375.1 | - | 12 - 13 | 1394 | 132 | ||
| BC012875.1 | - | 12 - 13 | 1341 | 3500318 | 132 | |
| CT010331.1 | - | 11 - 12 | 1272 | 132 | ||
| U65986.1 | - | 12 - 13 | 1406 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa11
Genomic Map