Assay Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008914.2 | NP_032940.1 | 1 - 2 | 220 | 134 | |
| GenBank mRNA | BC066000.1 | - | 1 - 2 | 191 | 5699179 | 134 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066000.1, NM_008914.2 transcripts.
Set Membership: |
|

Ppp3cb
Genomic Map