Assay Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008914.2 | NP_032940.1 | 7 - 8 | 1022 | 112 | |
| GenBank mRNA | AB221678.1 | - | 7 - 8 | 889 | 112 | |
| AK135203.1 | - | 7 - 8 | 836 | 112 | ||
| BC037668.1 | - | 7 - 8 | 845 | 112 | ||
| BC066000.1 | - | 7 - 8 | 993 | 5699179 | 112 | |
| M81483.1 | - | 7 - 8 | 859 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp3cb
Genomic Map