Assay Details
| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243041.1 | NP_001229970.1 | 9 - 10 | 978 | 64 | |
| NM_134079.4 | NP_598840.1 | 9 - 10 | 935 | 64 | ||
| GenBank mRNA | AK050135.1 | - | 3 - 4 | 238 | 64 | |
| AK151863.1 | - | 9 - 10 | 980 | 64 | ||
| AK160264.1 | - | 9 - 10 | 932 | 64 | ||
| AY540996.1 | - | 9 - 10 | 877 | 64 | ||
| AY540997.1 | - | 8 - 9 | 822 | 64 | ||
| BC009659.1 | - | 9 - 10 | 900 | 3485959 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adk
Genomic Map