Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005373.mm.1 |
NR_034153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005373.mm.1 |
NR_034153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005373.mm.1 |
TC1700002775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005379.mm.1 |
NM_026712.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005379.mm.1 |
NM_026712.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005379.mm.1 |
NR_034153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005379.mm.1 |
NR_034153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005379.mm.1 |
TC1700002775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005378.mm.1 |
NM_026712.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005378.mm.1 |
NM_026712.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005378.mm.1 |
NR_034153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005378.mm.1 |
NR_034153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005378.mm.1 |
TC1700002775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005376.mm.1 |
NM_026712.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005376.mm.1 |
NM_026712.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005376.mm.1 |
NR_034153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005376.mm.1 |
NR_034153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005376.mm.1 |
TC1700002775.mm.1 |
MTA TC |