Assay Details
| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177331.4 | NP_796305.3 | 1 - 2 | 198 | 75 | |
| GenBank mRNA | AK030995.1 | - | 1 - 2 | 208 | 75 | |
| AK031077.1 | - | 1 - 2 | 177 | 75 | ||
| BC090653.1 | - | 1 - 2 | 198 | 30659763 | 75 | |
| BC138157.1 | - | 1 - 2 | 85 | 8861176 | 75 | |
| BC138158.1 | - | 1 - 2 | 85 | 8861177 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030995.1, AK031077.1, BC090653.1, BC138157.1, BC138158.1, NM_177331.4 transcripts.
Set Membership: |
|

Gen1
Genomic Map