Assay Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027636.1 | 8 - 9 | 776 | 74 | ||
| GenBank mRNA | AK016201.1 | - | 8 - 9 | 771 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Daw1
Genomic Map