Assay Details
| Gene Symbol: Pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013737.5 | NP_038765.2 | 9 - 10 | 1236 | 73 | |
| GenBank mRNA | AK005158.1 | - | 9 - 10 | 973 | 73 | |
| AK005210.1 | - | 9 - 10 | 1030 | 73 | ||
| AK051454.1 | - | 9 - 10 | 1237 | 73 | ||
| BC010726.1 | - | 9 - 10 | 992 | 3707891 | 73 | |
| U34277.1 | - | 9 - 10 | 982 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g7
Genomic Map