Assay Details
| Gene Symbol: Pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013737.5 | NP_038765.2 | 2 - 3 | 471 | 75 | |
| GenBank mRNA | AK005158.1 | - | 2 - 3 | 208 | 75 | |
| AK005210.1 | - | 2 - 3 | 265 | 75 | ||
| AK051454.1 | - | 2 - 3 | 472 | 75 | ||
| AK155678.1 | - | 3 - 4 | 322 | 75 | ||
| BC010726.1 | - | 2 - 3 | 227 | 3707891 | 75 | |
| U34277.1 | - | 2 - 3 | 217 | 75 | ||
More Information
Set Membership: |
|

Pla2g7
Genomic Map