Assay Details
| Gene Symbol: Pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013737.5 | NP_038765.2 | 7 - 8 | 1032 | 102 | |
| GenBank mRNA | AK005158.1 | - | 7 - 8 | 769 | 102 | |
| AK051454.1 | - | 7 - 8 | 1033 | 102 | ||
| BC010726.1 | - | 7 - 8 | 788 | 3707891 | 102 | |
| U34277.1 | - | 7 - 8 | 778 | 102 | ||
More Information
Set Membership: |
|

Pla2g7
Genomic Map