Assay Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023162.4 | NP_075651.1 | 2 - 3 | 380 | 72 | |
| GenBank mRNA | AF230339.1 | - | 2 - 3 | 253 | 72 | |
| AK003714.1 | - | 2 - 3 | 381 | 72 | ||
| AK005976.1 | - | 3 - 4 | 353 | 72 | ||
| AK019152.1 | - | 3 - 4 | 357 | 72 | ||
| BC043016.1 | - | 3 - 4 | 317 | 5683859 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Znrd1
Genomic Map