Assay Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277863.1 | NP_001264792.1 | 2 - 3 | 357 | 123 | |
| NM_010021.5 | NP_034151.3 | 2 - 3 | 357 | 123 | ||
| GenBank mRNA | AK014854.1 | - | 2 - 3 | 337 | 123 | |
| AK033183.1 | - | 2 - 3 | 337 | 123 | ||
| AK033316.1 | - | 2 - 3 | 337 | 123 | ||
| AK160855.1 | - | 2 - 3 | 216 | 123 | ||
| BC099940.1 | - | 2 - 3 | 317 | 30048233 | 123 | |
| U38690.1 | - | 2 - 3 | 472 | 123 | ||
| U46694.1 | - | 2 - 3 | 253 | 123 | ||
| X95724.1 | - | 2 - 3 | 246 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160855.1 transcripts.
Set Membership: |
|

Dazl
Genomic Map