Assay Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277863.1 | NP_001264792.1 | 4 - 5 | 511 | 71 | |
| NM_010021.5 | NP_034151.3 | 4 - 5 | 511 | 71 | ||
| GenBank mRNA | AK014854.1 | - | 4 - 5 | 491 | 71 | |
| AK033183.1 | - | 4 - 5 | 491 | 71 | ||
| AK033316.1 | - | 4 - 5 | 491 | 71 | ||
| AK160855.1 | - | 4 - 5 | 370 | 71 | ||
| BC099940.1 | - | 4 - 5 | 471 | 30048233 | 71 | |
| U38690.1 | - | 4 - 5 | 626 | 71 | ||
| U46694.1 | - | 4 - 5 | 407 | 71 | ||
| X95724.1 | - | 4 - 5 | 400 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dazl
Genomic Map