Assay Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK160855.1 | - | 1 - 2 | 73 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160855.1 transcripts.
Set Membership: |
|

Dazl
Genomic Map