Assay Details
| Gene Symbol: Kat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190846.1 | NP_001177775.1 | 16 - 17 | 1968 | 64 | |
| NM_020005.4 | NP_064389.2 | 16 - 17 | 2522 | 64 | ||
| GenBank mRNA | AF254442.1 | - | 16 - 17 | 2163 | 64 | |
| AK030070.1 | - | 16 - 17 | 1969 | 64 | ||
| AK132486.1 | - | 16 - 17 | 2223 | 64 | ||
| AK156290.1 | - | 16 - 17 | 2522 | 64 | ||
| BC019722.1 | - | 4 - 5 | 328 | 64 | ||
| BC082581.1 | - | 16 - 17 | 2178 | 30653109 | 64 | |
| BC138195.1 | - | 16 - 17 | 2190 | 8861214 | 64 | |
| BC145896.1 | - | 16 - 17 | 2190 | 40131199 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat2b
Genomic Map