Assay Details
| Gene Symbol: Kat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190846.1 | NP_001177775.1 | 1 - 2 | 52 | 66 | |
| GenBank mRNA | AK030070.1 | - | 1 - 2 | 53 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190846.1 transcripts.
Set Membership: |
|

Kat2b
Genomic Map