Assay Details
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161722.1 | NP_001155194.1 | 5 - 6 | 963 | 77 | |
| NM_001161723.1 | NP_001155195.1 | 5 - 6 | 957 | 77 | ||
| NM_011549.3 | NP_035679.3 | 4 - 5 | 904 | 77 | ||
| GenBank mRNA | AF079095.1 | - | 4 - 5 | 885 | 77 | |
| AK078755.1 | - | 5 - 6 | 895 | 77 | ||
| AK146016.1 | - | 4 - 5 | 929 | 77 | ||
| AK154971.1 | - | 5 - 6 | 961 | 77 | ||
| BC064789.1 | - | 4 - 5 | 904 | 30429545 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfeb
Genomic Map