Assay Details
| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009085.2 | NP_033111.2 | 3 - 4 | 328 | 58 | |
| GenBank mRNA | AK132114.1 | - | 3 - 4 | 270 | 58 | |
| AK146240.1 | - | 3 - 4 | 278 | 58 | ||
| BC130018.1 | - | 3 - 4 | 329 | 40140532 | 58 | |
| BY267923.1 | - | 3 - 4 | 328 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr1c
Genomic Map