Assay Details
| Gene Symbol: Sh3gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252471.1 | NP_001239400.1 | 6 - 7 | 796 | 84 | |
| NM_013664.3 | NP_038692.1 | 6 - 7 | 796 | 84 | ||
| GenBank mRNA | AK154836.1 | - | 6 - 7 | 795 | 84 | |
| AK162715.1 | - | 6 - 7 | 724 | 84 | ||
| AK167505.1 | - | 6 - 7 | 738 | 84 | ||
| BC138602.1 | - | 6 - 7 | 735 | 8861622 | 84 | |
| BC138603.1 | - | 6 - 7 | 735 | 8861623 | 84 | |
| BC145345.1 | - | 6 - 7 | 735 | 84 | ||
| U58885.1 | - | 6 - 7 | 735 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3gl1
Genomic Map