Assay Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032572.1 | - | 11 - 12 | 3090 | 54 | |
| AK143616.1 | - | 15 - 16 | 2021 | 54 | ||
| AK146918.1 | - | 15 - 16 | 1998 | 54 | ||
| FN687734.2 | - | 15 - 16 | 1747 | 54 | ||
More Information
Set Membership: |
|

Hdgfrp2
Genomic Map