Assay Details
| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026768.3 | NP_081044.1 | 3 - 4 | 267 | 68 | |
| GenBank mRNA | AB049953.1 | - | 3 - 4 | 276 | 68 | |
| AK003444.1 | - | 3 - 4 | 268 | 68 | ||
| AK028226.1 | - | 3 - 4 | 269 | 68 | ||
| AK172171.1 | - | 3 - 4 | 262 | 68 | ||
| BC025456.1 | - | 1 - 2 | 63 | 68 | ||
| BC058959.1 | - | 3 - 4 | 255 | 6817698 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps18a
Genomic Map