Assay Details
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010321.1 | NP_034451.1 | 4 - 5 | 607 | 234 | |
| GenBank mRNA | AK007398.1 | - | 4 - 5 | 433 | 234 | |
| BC014283.1 | - | 4 - 5 | 606 | 4210236 | 234 | |
| BC087960.1 | - | 4 - 5 | 607 | 6766468 | 234 | |
| D89664.1 | - | 4 - 5 | 607 | 234 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnmt
Genomic Map