Assay Details
| Gene Symbol: Trem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272078.1 | NP_001259007.1 | 3 - 4 | 643 | 59 | |
| GenBank mRNA | AK155163.1 | - | 3 - 4 | 557 | 59 | |
| AY187009.1 | - | 3 - 4 | 510 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trem2
Genomic Map