Assay Details
| Gene Symbol: Ninl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207204.2 | NP_997087.2 | 13 - 14 | 1849 | 73 | |
| GenBank mRNA | AK129253.1 | - | 13 - 14 | 1782 | 73 | |
| BC067071.1 | - | 13 - 14 | 1783 | 30615484 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ninl
Genomic Map