Assay Details
| Gene Symbol: Atat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142744.1 | NP_001136216.1 | 11 - 12 | 1083 | 113 | |
| GenBank mRNA | AK011750.2 | - | 11 - 12 | 1059 | 113 | |
| AK011838.1 | - | 11 - 12 | 1074 | 113 | ||
| BC028847.1 | - | 11 - 12 | 1062 | 4007862 | 113 | |
| BC030669.1 | - | 10 - 11 | 992 | 1246856 | 113 | |
| BQ748472.1 | - | 2 - 3 | 119 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atat1
Genomic Map