Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 2 - 3 | 198 | 150 | |
| GenBank mRNA | AK044203.1 | - | 2 - 3 | 171 | 150 | |
| AK137158.1 | - | 2 - 3 | 142 | 150 | ||
| AK140375.1 | - | 2 - 3 | 171 | 150 | ||
| AK170850.1 | - | 2 - 3 | 171 | 150 | ||
| BC012875.1 | - | 2 - 3 | 118 | 3500318 | 150 | |
| BY723474.1 | - | 2 - 3 | 199 | 150 | ||
| U65986.1 | - | 2 - 3 | 183 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK140375.1, BC012875.1, NM_013469.2, U65986.1 transcripts.
Set Membership: |
|

Anxa11
Genomic Map