Assay Details
| Gene Symbol: Daam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008231.2 | NP_001008232.2 | 22 - 23 | 2957 | 85 | |
| GenBank mRNA | AK009382.1 | - | 7 - 8 | 779 | 85 | |
| AK122266.1 | - | 20 - 21 | 5875 | 85 | ||
| AY426536.1 | - | 22 - 23 | 2986 | 85 | ||
| BC037720.1 | - | 6 - 7 | 678 | 85 | ||
More Information
Set Membership: |
|

Daam2
Genomic Map