Assay Details
| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026987.2 | NP_081263.2 | 6 - 7 | 1266 | 65 | |
| GenBank mRNA | AK122314.2 | - | 6 - 7 | 1229 | 65 | |
| BC046781.1 | - | 6 - 7 | 1219 | 5716182 | 65 | |
| BU054791.1 | - | 1 - 2 | 99 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dhx16
Genomic Map