Assay Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025408.2 | NP_079684.2 | 3 - 4 | 347 | 111 | |
| GenBank mRNA | AK011668.1 | - | 3 - 4 | 328 | 111 | |
| AK017361.1 | - | 252 | 111 | |||
| AK080977.1 | - | 3 - 4 | 325 | 111 | ||
| AK150629.1 | - | 3 - 4 | 308 | 111 | ||
| AK162403.1 | - | 3 - 4 | 363 | 111 | ||
| BC023924.1 | - | 3 - 4 | 351 | 5324078 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acer3
Genomic Map