Assay Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025408.2 | NP_079684.2 | 8 - 9 | 676 | 157 | |
| GenBank mRNA | AK004287.1 | - | 8 - 9 | 677 | 157 | |
| AK011668.1 | - | 8 - 9 | 657 | 157 | ||
| AK017361.1 | - | 581 | 157 | |||
| AK080977.1 | - | 8 - 9 | 654 | 157 | ||
| AK150629.1 | - | 8 - 9 | 637 | 157 | ||
| AK162403.1 | - | 8 - 9 | 692 | 157 | ||
| BC023924.1 | - | 9 - 10 | 844 | 5324078 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acer3
Genomic Map