Assay Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242363.2 | NP_001229292.1 | 11 - 12 | 2864 | 70 | |
| NM_001242364.2 | NP_001229293.1 | 10 - 11 | 2815 | 70 | ||
| NM_001242365.2 | NP_001229294.1 | 10 - 11 | 2221 | 70 | ||
| NM_199009.3 | NP_950174.1 | 10 - 11 | 2773 | 70 | ||
| GenBank mRNA | AK085350.1 | - | 10 - 11 | 2769 | 70 | |
| AK154173.1 | - | 11 - 12 | 2808 | 70 | ||
| AK155273.1 | - | 11 - 12 | 2809 | 70 | ||
| AK159716.1 | - | 10 - 11 | 2176 | 70 | ||
| AK169996.1 | - | 11 - 12 | 2846 | 70 | ||
| BC018464.1 | - | 10 - 11 | 2732 | 70 | ||
| BC019738.1 | - | 2 - 3 | 647 | 70 | ||
| BC021532.1 | - | 8 - 9 | 2142 | 70 | ||
| BC043717.1 | - | 9 - 10 | 2289 | 70 | ||
| BC063093.1 | - | 10 - 11 | 2730 | 6332038 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam160a2
Genomic Map