Assay Details
| Gene Symbol: P4ha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177161.4 | NP_796135.3 | 2 - 3 | 351 | 62 | |
| GenBank mRNA | AK086780.1 | - | 2 - 3 | 332 | 62 | |
| AY313449.1 | - | 2 - 3 | 351 | 62 | ||
| BC082538.1 | - | 2 - 3 | 393 | 5701368 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC082538.1 transcripts.
Set Membership: |
|

P4ha3
Genomic Map