Assay Details
Gene Symbol: Olfr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168503.1 | NP_001161975.1 | 1 - 2 | 158 | 89 | |
GenBank mRNA | AK036356.1 | - | 1 - 2 | 159 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK036356.1, NM_001168503.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
