Assay Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008034.3 | NP_032060.2 | 2 - 3 | 327 | 95 | |
| GenBank mRNA | AF096319.1 | - | 2 - 3 | 222 | 95 | |
| AK002275.1 | - | 2 - 3 | 325 | 95 | ||
| CD241494.1 | - | 2 - 3 | 340 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF096319.1, NM_008034.3 transcripts.
Set Membership: |
|

Folr1
Genomic Map