Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 16 - 17 | 1744 | 69 | |
| GenBank mRNA | AB476647.1 | - | 15 - 16 | 1620 | 69 | |
| AB476648.1 | - | 16 - 17 | 1683 | 69 | ||
| AK010773.1 | - | 15 - 16 | 1665 | 69 | ||
| AK131109.1 | - | 1678 | 69 | |||
| AK155125.1 | - | 16 - 17 | 1734 | 69 | ||
| AK157410.1 | - | 16 - 17 | 1724 | 69 | ||
| AK158186.1 | - | 15 - 16 | 1853 | 69 | ||
| BC060054.1 | - | 15 - 16 | 2240 | 69 | ||
| BP767155.1 | - | 2 - 3 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map