Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 13 - 14 | 1438 | 85 | |
| GenBank mRNA | AB476647.1 | - | 12 - 13 | 1314 | 85 | |
| AB476648.1 | - | 13 - 14 | 1377 | 85 | ||
| AK010773.1 | - | 12 - 13 | 1359 | 85 | ||
| AK155125.1 | - | 13 - 14 | 1428 | 85 | ||
| AK157410.1 | - | 13 - 14 | 1417 | 85 | ||
| AK158186.1 | - | 12 - 13 | 1552 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map