Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 1 - 2 | 184 | 68 | |
| GenBank mRNA | AB476647.1 | - | 1 - 2 | 123 | 68 | |
| AB476648.1 | - | 1 - 2 | 123 | 68 | ||
| AK010773.1 | - | 1 - 2 | 167 | 68 | ||
| AK131109.1 | - | 155 | 68 | |||
| AK155125.1 | - | 1 - 2 | 173 | 68 | ||
| AK157410.1 | - | 1 - 2 | 163 | 68 | ||
| AK158186.1 | - | 1 - 2 | 124 | 68 | ||
| BC060054.1 | - | 1 - 2 | 347 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map