Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 2 - 3 | 286 | 75 | |
| GenBank mRNA | AB476647.1 | - | 2 - 3 | 225 | 75 | |
| AB476648.1 | - | 2 - 3 | 225 | 75 | ||
| AK010773.1 | - | 2 - 3 | 269 | 75 | ||
| AK131109.1 | - | 257 | 75 | |||
| AK155125.1 | - | 2 - 3 | 275 | 75 | ||
| AK157410.1 | - | 2 - 3 | 265 | 75 | ||
| AK158186.1 | - | 2 - 3 | 226 | 75 | ||
| BC060054.1 | - | 2 - 3 | 449 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map