Assay Details
| Gene Symbol: Arl6ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019419.2 | NP_062292.1 | 2 - 3 | 287 | 65 | |
| GenBank mRNA | AF133669.1 | - | 2 - 3 | 232 | 65 | |
| AF172088.1 | - | 2 - 3 | 273 | 65 | ||
| AF223953.1 | - | 2 - 3 | 225 | 65 | ||
| AK090001.1 | - | 2 - 3 | 239 | 65 | ||
| AK150042.1 | - | 2 - 3 | 240 | 65 | ||
| AK150507.1 | - | 2 - 3 | 238 | 65 | ||
| AK151272.1 | - | 2 - 3 | 237 | 65 | ||
| AK151507.1 | - | 2 - 3 | 237 | 65 | ||
| AK152565.1 | - | 2 - 3 | 237 | 65 | ||
| AK152885.1 | - | 2 - 3 | 234 | 65 | ||
| AK154355.1 | - | 2 - 3 | 259 | 65 | ||
| AK154644.1 | - | 2 - 3 | 240 | 65 | ||
| AK163655.1 | - | 2 - 3 | 237 | 65 | ||
| AK167872.1 | - | 2 - 3 | 242 | 65 | ||
| AK167888.1 | - | 2 - 3 | 290 | 65 | ||
| AK167912.1 | - | 2 - 3 | 237 | 65 | ||
| AK168172.1 | - | 2 - 3 | 240 | 65 | ||
| AK168245.1 | - | 2 - 3 | 239 | 65 | ||
| AK169191.1 | - | 2 - 3 | 237 | 65 | ||
| AK176958.1 | - | 2 - 3 | 148 | 65 | ||
| BC010196.1 | - | 2 - 3 | 227 | 3157834 | 65 | |
| BC046792.1 | - | 2 - 3 | 210 | 5717527 | 65 | |
| EF437380.1 | - | 2 - 3 | 171 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arl6ip1
Genomic Map