Assay Details
| Gene Symbol: Eif4g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013507.3 | NP_038535.2 | 13 - 14 | 1626 | 83 | |
| GenBank mRNA | AK141512.1 | - | 6 - 7 | 696 | 83 | |
| AK144309.1 | - | 13 - 14 | 1600 | 83 | ||
| AK150745.1 | - | 13 - 14 | 1603 | 83 | ||
| AK151637.1 | - | 13 - 14 | 1603 | 83 | ||
| BC040391.1 | - | 12 - 13 | 1315 | 4018962 | 83 | |
| BC043034.1 | - | 12 - 13 | 1306 | 5696921 | 83 | |
| BC056387.1 | - | 13 - 14 | 1590 | 6854755 | 83 | |
| BC064810.1 | - | 12 - 13 | 1359 | 6825556 | 83 | |
| BC092521.1 | - | 13 - 14 | 1581 | 30649919 | 83 | |
| BC127064.1 | - | 13 - 14 | 1626 | 40109978 | 83 | |
| U76112.1 | - | 13 - 14 | 1600 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4g2
Genomic Map