Assay Details
| Gene Symbol: Eif4g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040131.2 | NP_001035221.1 | 6 - 7 | 811 | 105 | |
| NM_013507.3 | NP_038535.2 | 6 - 7 | 811 | 105 | ||
| GenBank mRNA | AK144309.1 | - | 6 - 7 | 785 | 105 | |
| AK150745.1 | - | 6 - 7 | 788 | 105 | ||
| AK151637.1 | - | 6 - 7 | 788 | 105 | ||
| BC039274.1 | - | 5 - 6 | 489 | 105 | ||
| BC040391.1 | - | 5 - 6 | 500 | 4018962 | 105 | |
| BC043034.1 | - | 5 - 6 | 491 | 5696921 | 105 | |
| BC056387.1 | - | 6 - 7 | 775 | 6854755 | 105 | |
| BC057673.1 | - | 7 - 8 | 663 | 4020759 | 105 | |
| BC064810.1 | - | 5 - 6 | 544 | 6825556 | 105 | |
| BC092521.1 | - | 6 - 7 | 766 | 30649919 | 105 | |
| BC127064.1 | - | 6 - 7 | 811 | 40109978 | 105 | |
| CK619644.1 | - | 4 - 5 | 401 | 105 | ||
| U63323.1 | - | 6 - 7 | 763 | 105 | ||
| U76112.1 | - | 6 - 7 | 785 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144309.1, AK150745.1, AK151637.1 transcripts.
Set Membership: |
|

Eif4g2
Genomic Map