Assay Details
| Gene Symbol: Eif4g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040131.2 | NP_001035221.1 | 8 - 9 | 1031 | 107 | |
| NM_013507.3 | NP_038535.2 | 8 - 9 | 1031 | 107 | ||
| GenBank mRNA | AK141512.1 | - | 1 - 2 | 101 | 107 | |
| AK144309.1 | - | 8 - 9 | 1005 | 107 | ||
| AK150745.1 | - | 8 - 9 | 1008 | 107 | ||
| AK151637.1 | - | 8 - 9 | 1008 | 107 | ||
| BC039274.1 | - | 7 - 8 | 709 | 107 | ||
| BC040391.1 | - | 7 - 8 | 720 | 4018962 | 107 | |
| BC043034.1 | - | 7 - 8 | 711 | 5696921 | 107 | |
| BC056387.1 | - | 8 - 9 | 995 | 6854755 | 107 | |
| BC057673.1 | - | 9 - 10 | 883 | 4020759 | 107 | |
| BC064810.1 | - | 7 - 8 | 764 | 6825556 | 107 | |
| BC092521.1 | - | 8 - 9 | 986 | 30649919 | 107 | |
| BC127064.1 | - | 8 - 9 | 1031 | 40109978 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4g2
Genomic Map