Assay Details
| Gene Symbol: Calca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033954.3 | NP_001029126.1 | 4 - 5 | 563 | 164 | |
| NM_001289444.1 | NP_001276373.1 | 4 - 5 | 587 | 164 | ||
| GenBank mRNA | AF330212.1 | - | 4 - 5 | 501 | 164 | |
| BC028771.1 | - | 4 - 5 | 496 | 1331673 | 164 | |
More Information
Updated Mapping Information
Set Membership: |
|

Calca
Genomic Map