Assay Details
| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025559.3 | NP_001020730.1 | 2 - 3 | 130 | 60 | |
| NM_001025560.2 | NP_001020731.1 | 2 - 3 | 250 | 60 | ||
| NM_011445.4 | NP_035575.1 | 2 - 3 | 158 | 60 | ||
| GenBank mRNA | AK030713.1 | - | 2 - 3 | 251 | 60 | |
| AK048143.1 | - | 2 - 3 | 131 | 60 | ||
| AK084290.1 | - | 2 - 3 | 289 | 60 | ||
| U32614.1 | - | 2 - 3 | 162 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030713.1, NM_001025559.3, NM_001025560.2, NM_011445.4, U32614.1 transcripts.
Set Membership: |
|

Sox6
Genomic Map