Assay Details
| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025560.2 | NP_001020731.1 | 7 - 8 | 1036 | 97 | |
| NM_001277326.1 | NP_001264255.1 | 6 - 7 | 891 | 97 | ||
| NM_001277328.1 | NP_001264257.1 | 5 - 6 | 779 | 97 | ||
| NM_011445.4 | NP_035575.1 | 7 - 8 | 944 | 97 | ||
| GenBank mRNA | AK029696.1 | - | 5 - 6 | 706 | 97 | |
| AK030713.1 | - | 7 - 8 | 1037 | 97 | ||
| AK084290.1 | - | 7 - 8 | 1075 | 97 | ||
| AK132409.1 | - | 6 - 7 | 857 | 97 | ||
| D61689.1 | - | 5 - 6 | 775 | 97 | ||
| U32614.1 | - | 7 - 8 | 948 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox6
Genomic Map