Assay Details
| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009667.3 | NP_033797.2 | 1 - 2 | 292 | 121 | |
| GenBank mRNA | AK133465.1 | - | 1 - 2 | 159 | 121 | |
| AK171276.1 | - | 1 - 2 | 109 | 121 | ||
| BC040366.1 | - | 1 - 2 | 292 | 4503933 | 121 | |
| BC056380.1 | - | 1 - 2 | 134 | 6851948 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133465.1, BC040366.1, BC056380.1, NM_009667.3 transcripts.
Set Membership: |
|

Ampd3
Genomic Map