Assay Details
| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276301.1 | NP_001263230.1 | 2 - 3 | 398 | 73 | |
| NM_009667.3 | NP_033797.2 | 2 - 3 | 517 | 73 | ||
| GenBank mRNA | AK133465.1 | - | 2 - 3 | 384 | 73 | |
| AK171276.1 | - | 2 - 3 | 334 | 73 | ||
| BC040366.1 | - | 2 - 3 | 517 | 4503933 | 73 | |
| BC056380.1 | - | 2 - 3 | 359 | 6851948 | 73 | |
| D85596.1 | - | 2 - 3 | 396 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ampd3
Genomic Map